4,4'-Dibromo-2-nitrobiphenyl ≥98.0%

Leverantör: TCI
Varning

D5189-1G D5189-5G
TCIAD5189-1GEA 885 SEK
TCIAD5189-1G TCIAD5189-5G
4,4'-Dibromo-2-nitrobiphenyl ≥98.0%
4,4'-Dibromo-2-nitrobiphenyl
Formel: C₁₂H₇Br₂NO₂
MDL Number: MFCD05854318
CAS nummer: 439797-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0aae851ce7eeaa40bd4f722c5c2d08b5f6be642e&vid=0aae851ce7eeaa40bd4f722c5c2d08b5f6be642e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR