(3,5-Dimethoxyphenyl)acetonitrile ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile
D5186-1G D5186-5G
TCIAD5186-1GEA 525 SEK
TCIAD5186-1G TCIAD5186-5G
(3,5-Dimethoxyphenyl)acetonitrile ≥98.0%
(3,5-Dimethoxyphenyl)acetonitrile
Formel: C₁₀H₁₁NO₂
MW: 177,2 g/mol
MDL Number: MFCD00016395
CAS nummer: 13388-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd9dd3a51f25306836f84d4ff536df16c06f8b7c&vid=dd9dd3a51f25306836f84d4ff536df16c06f8b7c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 58.0 deg-C