1,2-Dimethyl-3H-benzo[e]indole ≥98.0%

Leverantör: TCI EUROPE

D5163-1G D5163-5G
TCIAD5163-1GEA 525 SEK
TCIAD5163-1G TCIAD5163-5G
1,2-Dimethyl-3H-benzo[e]indole ≥98.0%
1,2-Dimethyl-3H-benzo[e]indole
Formel: C₁₄H₁₃N
MW: 195,27 g/mol
MDL Number: MFCD00850046
CAS nummer: 55970-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e120aa7682a3aa28bb42ca757b126f699f775da8&vid=e120aa7682a3aa28bb42ca757b126f699f775da8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR