2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole ≥98.0%

Leverantör: TCI EUROPE
Varning

D5162-1G D5162-5G
TCIAD5162-1GEA 525 SEK
TCIAD5162-1G TCIAD5162-5G
2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole ≥98.0%
2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole
Formel: C₁₆H₁₇N
MW: 223,32 g/mol
CAS nummer: 344928-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d56410b705cb8662b2083babd3ab78093a91a0b&vid=4d56410b705cb8662b2083babd3ab78093a91a0b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR