5,6-Dimethoxyindole ≥98.0%

Leverantör: TCI EUROPE
Varning

D5148-200MG D5148-1G
TCIAD5148-200MGEA 590 SEK
TCIAD5148-200MG TCIAD5148-1G
5,6-Dimethoxyindole ≥98.0%
5,6-Dimethoxyindole
Formel: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Kokpunkt: 198°C
MDL Number: MFCD00005675
CAS nummer: 14430-23-0
EINECS: 238-402-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5684f8f4fee2a8fde8dca01fe4807fa306ccde2&vid=a5684f8f4fee2a8fde8dca01fe4807fa306ccde2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light gray to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 153.0 to 157.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR