2,3-Difluoroisonicotinic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2,3-Difluoropyridine-4-carboxylic acid

D5131-1G D5131-5G
TCIAD5131-1GEA 285 SEK
TCIAD5131-1G TCIAD5131-5G
2,3-Difluoroisonicotinic acid ≥98.0% (by GC, titration analysis)
2,3-Difluoroisonicotinic acid
Formel: C₆H₃F₂NO₂
MW: 159,09 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09031174
CAS nummer: 851386-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35e4ca929798cfaf503da9d44dba2f016eafc639&vid=35e4ca929798cfaf503da9d44dba2f016eafc639&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR