α-Chloro-3,4-difluorotoulene ≥96.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 1-(Chloromethyl)-3,4-difluorobenzene, 3,4-Difluorobenzyl chloride, 4-(Chloromethyl)-1,2-difluorobenzene
D5117-5G D5117-25G
TCIAD5117-5GEA 510 SEK
TCIAD5117-5G TCIAD5117-25G
α-Chloro-3,4-difluorotoulene ≥96.0%
α-Chloro-3,4-difluorotoulene
Formel: C₇H₅ClF₂
MW: 162,57 g/mol
Kokpunkt: 80°C
MDL Number: MFCD00236218
CAS nummer: 698-80-6
UN: 3265
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %