3',5'-Dichloroacetophenone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(3,5-Dichlorophenyl)ethanone

D5111-1G D5111-5G
TCIAD5111-1GEA 365 SEK
TCIAD5111-1G TCIAD5111-5G
3',5'-Dichloroacetophenone ≥98.0%
3',5'-Dichloroacetophenone
Formel: C₈H₆Cl₂O
MW: 189,04 g/mol
Kokpunkt: 135°C
MDL Number: MFCD00045189
CAS nummer: 14401-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32537fe3611ee1d041e182865e4bc753889a34d5&vid=32537fe3611ee1d041e182865e4bc753889a34d5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR