3,5-Dibromo-2-methoxypyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

D5099-1G D5099-5G
TCIAD5099-1GEA 295 SEK
TCIAD5099-1G TCIAD5099-5G
3,5-Dibromo-2-methoxypyridine ≥98.0%
3,5-Dibromo-2-methoxypyridine
Formel: C₆H₅Br₂NO
MW: 266,9200 g/mol
Kokpunkt: 120°C
MDL Number: MFCD05663709
CAS nummer: 13472-60-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 and le 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR