2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,4-Dimethylphenylboronic acid Pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene
D5085-1G
D5085-5G
TCIAD5085-1GEA
575
SEK
TCIAD5085-1G
TCIAD5085-5G
2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel:
C₁₄H₂₁BO₂ MW: 232,13 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD12405519 CAS nummer: 401797-00-0 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...