Diisopropylamine dichloroacetate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Diisopropylammonium dichloroacetate

D5074-1G D5074-5G
TCIAD5074-1GEA 750 SEK
TCIAD5074-1G TCIAD5074-5G
Diisopropylamine dichloroacetate ≥98.0%
Diisopropylamine dichloroacetate
Formel: C₈H₁₇Cl₂NO₂
MW: 230.129 g/mol
MDL Number: MFCD00868288
CAS nummer: 660-27-5
EINECS: 211-538-2
Merck Index: 14,03197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cadee42570775224f2819376f9650e519841e89&vid=8cadee42570775224f2819376f9650e519841e89&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR