Decahydroisoquinoline cis- and trans-mixture ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Decahydroisoquinoline (cis- and trans- mixture, predominantly cis-isomer)
D5067-5ML
D5067-25ML
TCIAD5067-5MLEA
1250
SEK
TCIAD5067-5ML
TCIAD5067-25ML
Decahydroisoquinoline cis- and trans-mixture ≥98.0%
Decahydroisoquinoline
Formel:
C₉H₁₇N Kokpunkt: 80°C Flampunkt: 79°C |
MDL Number:
MFCD00012096 CAS nummer: 6329-61-9 EINECS: 228-702-4 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...