2,7-Dibromo-9-hexylcarbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,7-Dibromo-9-hexyl-9H-carbazole
D5055-1G
D5055-5G
TCIAD5055-1GEA
565
SEK
TCIAD5055-1G
TCIAD5055-5G
2,7-Dibromo-9-hexylcarbazole ≥98.0%
2,7-Dibromo-9-hexylcarbazole
Formel:
C₁₈H₁₉Br₂N MW: 409,17 g/mol |
MDL Number:
MFCD29089359 CAS nummer: 654676-12-7 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...