9,9-Diphenyl-9H-9-silafluorene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 5,5-Diphenyl-5H-dibenzo[b,d]silole

D5034-200MG D5034-1G
TCIAD5034-200MGEA 805 SEK
TCIAD5034-200MG TCIAD5034-1G
9,9-Diphenyl-9H-9-silafluorene ≥98.0%
9,9-Diphenyl-9H-9-silafluorene
Formel: C₂₄H₁₈Si
MW: 334,49 g/mol
CAS nummer: 5550-08-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR