2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide ≥98.0%

Leverantör: TCI

Synonyms: 1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione

D5021-200MG D5021-1G
TCIAD5021-200MGEA 685 SEK
TCIAD5021-200MG TCIAD5021-1G
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide ≥98.0%
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide
Formel: C₁₄H₁₇Br₂NO₂S
MDL Number: MFCD23703120
CAS nummer: 1231160-83-0

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 95.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR