4,5-Dibromoveratrole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,2-Dibromo-4,5-dimethoxybenzene, 4,5-Dibromoveratrole
D5019-5G
D5019-25G
TCIAD5019-5GEA
485
SEK
TCIAD5019-5G
TCIAD5019-25G
4,5-Dibromoveratrole ≥98.0%
4,5-Dibromoveratrole
Formel:
C₈H₈Br₂O₂ MW: 295,96 g/mol |
MDL Number:
MFCD00014894 CAS nummer: 37895-73-1 EINECS: 253-705-2 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...