1,5-Diiodo-2,4-dimethylbenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4,6-Diiodo-m-xylene
D5005-1G
D5005-5G
TCIAD5005-1GEA
375
SEK
TCIAD5005-1G
TCIAD5005-5G
1,5-Diiodo-2,4-dimethylbenzene ≥97.0%
1,5-Diiodo-2,4-dimethylbenzene
Formel:
C₈H₈I₂ MW: 357,96 g/mol |
CAS nummer:
4102-50-5 |
Specifikation testresultat
Purity (GC) | min. 97.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...