Dimethyl-2-bromoterephthalate ≥98.0%

Leverantör: TCI

Synonyms: Dimethyl 2-bromoterephthalate, Bromoterephthalic acid dimethyl ester, Dimethyl Bromoterephthalate

D5003-5G D5003-25G
TCIAD5003-5GEA 1200 SEK
TCIAD5003-5G TCIAD5003-25G
Dimethyl-2-bromoterephthalate ≥98.0%
Dimethyl-2-bromoterephthalate
Formel: C₁₀H₉BrO₄
MDL Number: MFCD00060638
CAS nummer: 18643-86-2
EINECS: 242-476-4

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 53.0 and le 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR