3,6-Dibromo-9-(p-tolyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

D5001-200MG D5001-1G
TCIAD5001-200MGEA 495 SEK
TCIAD5001-200MG TCIAD5001-1G
3,6-Dibromo-9-(p-tolyl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(p-tolyl)-9H-carbazole
Formel: C₁₉H₁₃Br₂N
MW: 415,13 g/mol
MDL Number: MFCD12024282
CAS nummer: 357437-74-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR