3,3''-Dibromo-1,1':3',1''-terphenyl ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 3,3''-Dibromo-m-terphenyl
D4999-200MG D4999-1G
TCIAD4999-200MGEA 655 SEK
TCIAD4999-200MG TCIAD4999-1G
3,3''-Dibromo-1,1':3',1''-terphenyl ≥97.0%
3,3''-Dibromo-1,1':3',1''-terphenyl
Formel: C₁₈H₁₂Br₂
MW: 388,10 g/mol
Kokpunkt: 225°C
MDL Number: MFCD28138089
CAS nummer: 95962-62-2
UN: 3152
ADR: 9,II
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %