1H,1H,7H-Perfluoroheptyl methacrylate ≥97.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Methacrylate, Methacrylic acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl ester
Stabilization: med 4-tert-Butylpyrokatekol
D4990-5G D4990-25G
TCIAD4990-5GEA 470 SEK
TCIAD4990-5G TCIAD4990-25G
1H,1H,7H-Perfluoroheptyl methacrylate ≥97.0% stabilised
1H,1H,7H-Perfluoroheptyl methacrylate
Formel: C₁₁H₈F₁₂O₂
MW: 400,17 g/mol
Kokpunkt: 112°C
Flampunkt: 104°C
Storage Temperature: Kylskåp
MDL Number: MFCD00080658
CAS nummer: 2261-99-6
EINECS: 218-863-9
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %