1,4-Dibromo-2,3-difluorobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

D4983-1G D4983-5G
TCIAD4983-1GEA 510 SEK
TCIAD4983-1G TCIAD4983-5G
1,4-Dibromo-2,3-difluorobenzene ≥98.0%
1,4-Dibromo-2,3-difluorobenzene
Formel: C₆H₂Br₂F₂
MW: 271,8849 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09835192
CAS nummer: 156682-52-9

Order Now

Specifikation testresultat

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR