9-(3,5-Dibromophenyl)carbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 9-(3,5-Dibromophenyl)-9H-carbazole

D4978-200MG D4978-1G
TCIAD4978-200MGEA 750 SEK
TCIAD4978-200MG TCIAD4978-1G
9-(3,5-Dibromophenyl)carbazole ≥98.0%
9-(3,5-Dibromophenyl)carbazole
Formel: C₁₈H₁₁Br₂N
MW: 401,10 g/mol
MDL Number: MFCD27939624
CAS nummer: 750573-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1613cad966c59e61a6f467410e67963ca3a5d89&vid=d1613cad966c59e61a6f467410e67963ca3a5d89&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR