Diethylcyanamide ≥98.0%

Leverantör: TCI
Fara

D4965-1G D4965-5G
TCIAD4965-1GEA 295 SEK
TCIAD4965-1G TCIAD4965-5G
Diethylcyanamide ≥98.0%
Diethylcyanamide
Formel: C₅H₁₀N₂
Kokpunkt: 188°C
Flampunkt: 69°C
MDL Number: MFCD00001769
CAS nummer: 617-83-4
EINECS: 210-532-7
UN: 3276
ADR: 6.1 III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR