α,α,α,2,5-Pentafluorotoluene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,5-Difluorobenzotrifluoride
D4933-1G
D4933-5G
TCIAD4933-1GEA
295
SEK
TCIAD4933-1G
TCIAD4933-5G
α,α,α,2,5-Pentafluorotoluene ≥98.0%
α,α,α,2,5-Pentafluorotoluene
Formel:
C₇H₃F₅ MW: 182,09 g/mol Kokpunkt: 108°C Smältpunkt: –14°C |
MDL Number:
MFCD00042494 CAS nummer: 393-38-4 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...