2,6-Dimethylbenzonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4927-5G
D4927-25G
TCIAD4927-5GEA
685
SEK
TCIAD4927-5G
TCIAD4927-25G
2,6-Dimethylbenzonitrile ≥98.0%
2,6-Dimethylbenzonitrile
Formel:
C₉H₉N MW: 131,18 g/mol Kokpunkt: 93°C |
MDL Number:
MFCD00037166 CAS nummer: 6575-13-9 EINECS: 229-503-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...