2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2,3-Difluoro-1-methoxy-4-[(trans-4-propylcyclohexyl)methoxy]benzene

D4909-1G D4909-5G
TCIAD4909-1GEA 575 SEK
TCIAD4909-1G TCIAD4909-5G
2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole ≥98.0%
2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole
Formel: C₁₇H₂₄F₂O₂
MW: 298,37 g/mol
CAS nummer: 1373116-00-7

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 50.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR