1,4-Dibromo-2-iodobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4906-1G
D4906-5G
TCIAD4906-1GEA
845
SEK
TCIAD4906-1G
TCIAD4906-5G
1,4-Dibromo-2-iodobenzene ≥98.0%
1,4-Dibromo-2-iodobenzene
Formel:
C₆H₃Br₂I MW: 361,80 g/mol Kokpunkt: 180°C |
MDL Number:
MFCD07778993 CAS nummer: 89284-52-6 |
Specifikation testresultat
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...