9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

D4903-1G D4903-5G
TCIAD4903-1GEA 1900 SEK
TCIAD4903-1G TCIAD4903-5G
9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole ≥98.0%
9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole
Formel: C₃₆H₂₄N₂
MW: 484,60 g/mol
MDL Number: MFCD29905037
CAS nummer: 57102-62-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 204.0 to 208.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR