4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
D4895-200MG
D4895-1G
TCIAD4895-200MGEA
830
SEK
TCIAD4895-200MG
TCIAD4895-1G
4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine
Formel:
C₂₂H₁₄Br₂N₂ MW: 466,18 g/mol |
MDL Number:
MFCD16619323 CAS nummer: 607740-08-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 197.0 to 201.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...