N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 4-[Di(4-biphenylyl)amino]phenylboronic acid Pinacol ester, 2-[4-[Di(4-biphenylyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-([1,1'-Biphenyl]-4-yl)-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-[1,1'-biphenyl]-4-amine

D4886-1G D4886-5G
TCIAD4886-1GEA 935 SEK
TCIAD4886-1G TCIAD4886-5G
N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formel: C₃₆H₃₄BNO₂
MW: 523,48 g/mol
MDL Number: MFCD29089353
CAS nummer: 952431-30-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR