2,6-Di(2H-1,2,3-triazol-4-yl)pyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

D4879-200MG D4879-1G
TCIAD4879-200MGEA 725 SEK
TCIAD4879-200MG TCIAD4879-1G
2,6-Di(2H-1,2,3-triazol-4-yl)pyridine ≥98.0%
2,6-Di(2H-1,2,3-triazol-4-yl)pyridine
Formel: C₉H₇N₇
MW: 213,20 g/mol
CAS nummer: 1542234-40-1

Order Now

Specifikation testresultat

Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR