3,3-(Diphenyl)acrylonitrile ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,3-Diphenylacrylonitrile

D4873-200MG D4873-1G
TCIAD4873-200MGEA 695 SEK
TCIAD4873-200MG TCIAD4873-1G
3,3-(Diphenyl)acrylonitrile ≥98.0%
3,3-(Diphenyl)acrylonitrile
Formel: C₁₅H₁₁N
MW: 205,26 g/mol
Kokpunkt: 131°C
MDL Number: MFCD00045630
CAS nummer: 3531-24-6

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR