3',5'-Dimethylacetophenone ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 1-(3,5-Dimethylphenyl)ethanone, 1-(3,5-Dimethylphenyl)ethan-1-one

D4868-1G D4868-5G
TCIAD4868-1GEA 445 SEK
TCIAD4868-1G TCIAD4868-5G
3',5'-Dimethylacetophenone ≥97.0%
3',5'-Dimethylacetophenone
Formel: C₁₀H₁₂O
MW: 148,2 g/mol
Kokpunkt: 237°C
MDL Number: MFCD01075693
CAS nummer: 5379-16-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR