2,4-Dimethoxybenzonitrile ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
D4854-5G
D4854-25G
TCIAD4854-5GEA
390
SEK
TCIAD4854-5G
TCIAD4854-25G
2,4-Dimethoxybenzonitrile ≥99.0%
2,4-Dimethoxybenzonitrile
Formel:
C₉H₉NO₂ MW: 163,18 g/mol |
MDL Number:
MFCD00001786 CAS nummer: 4107-65-7 EINECS: 223-886-2 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...