5,6-Dibromo-1-hexene ≥96.0%
Leverantör: TCI EUROPE
Varning
D4844-1G D4844-5G
TCIAD4844-1GEA 1050 SEK
TCIAD4844-1G TCIAD4844-5G
5,6-Dibromo-1-hexene ≥96.0%
5,6-Dibromo-1-hexene
Formel: C₆H₁₀Br₂
MW: 241,95 g/mol
Kokpunkt: 55°C
Storage Temperature: Kylskåp
MDL Number: MFCD00041698
CAS nummer: 4285-48-7
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %