2,8-Diiododibenzofuran ≥98.0%
Leverantör: TCI EUROPE
D4835-200MG
D4835-1G
TCIAD4835-200MGEA
685
SEK
TCIAD4835-200MG
TCIAD4835-1G
2,8-Diiododibenzofuran ≥98.0%
2,8-Diiododibenzofuran
Formel:
C₁₂H₆I₂O MW: 419,99 g/mol |
MDL Number:
MFCD00094363 CAS nummer: 5943-11-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...