3,6-Dimethoxy-9H-carbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
D4833-200MG
D4833-1G
TCIAD4833-200MGEA
815
SEK
TCIAD4833-200MG
TCIAD4833-1G
3,6-Dimethoxy-9H-carbazole ≥98.0%
3,6-Dimethoxy-9H-carbazole
Formel:
C₁₄H₁₃NO₂ MW: 227,26 g/mol |
MDL Number:
MFCD00626046 CAS nummer: 57103-01-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...