3,4-Diacetyl-2,5-hexanedione ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Bis(acetylacetone), 1,1,2,2-Tetraacetylethane, 3,4-Diacetylhexane-2,5-dione

D4826-200MG D4826-1G
TCIAD4826-200MGEA 1350 SEK
TCIAD4826-200MG TCIAD4826-1G
3,4-Diacetyl-2,5-hexanedione ≥98.0%
3,4-Diacetyl-2,5-hexanedione
Formel: C₁₀H₁₄O₄
MW: 198,22 g/mol
Kokpunkt: 122°C
MDL Number: MFCD00031531
CAS nummer: 5027-32-7

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 191.0 to 195.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR