2,8-Dibromodibenzofuran ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,8-Dibromodibenzo[b,d]furan
D4821-1G
D4821-5G
TCIAD4821-1GEA
670
SEK
TCIAD4821-1G
TCIAD4821-5G
2,8-Dibromodibenzofuran ≥98.0%
2,8-Dibromodibenzofuran
Formel:
C₁₂H₆Br₂O MW: 325,99 g/mol |
MDL Number:
MFCD00093683 CAS nummer: 10016-52-1 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 193.0 to 197.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...