2'-Hydroxy-4',5'-dimethylacetophenone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(2-Hydroxy-4,5-dimethylphenyl)ethan-1-one
D4812-1G
D4812-5G
TCIAD4812-1GEA
430
SEK
TCIAD4812-1G
TCIAD4812-5G
2'-Hydroxy-4',5'-dimethylacetophenone ≥98.0%
2'-Hydroxy-4',5'-dimethylacetophenone
Formel:
C₁₀H₁₂O₂ MW: 164,21 g/mol Kokpunkt: 144°C |
MDL Number:
MFCD00002306 CAS nummer: 36436-65-4 EINECS: 253-035-0 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...