1,4-Difluoro-2-methyl-5-nitrobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,5-Difluoro-4-nitrotoluene

D4806-1G D4806-5G
TCIAD4806-1GEA 485 SEK
TCIAD4806-1G TCIAD4806-5G
1,4-Difluoro-2-methyl-5-nitrobenzene ≥98.0%
1,4-Difluoro-2-methyl-5-nitrobenzene
Formel: C₇H₅F₂NO₂
MW: 173,1169 g/mol
MDL Number: MFCD06200905
CAS nummer: 141412-60-4

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR