11,12-Dihydroindolo[2,3-a]carbazole ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
D4803-200MG
D4803-1G
TCIAD4803-200MGEA
350
SEK
TCIAD4803-200MG
TCIAD4803-1G
11,12-Dihydroindolo[2,3-a]carbazole ≥98.0% (by HPLC, total nitrogen)
11,12-Dihydroindolo[2,3-a]carbazole
Formel:
C₁₈H₁₂N₂ MW: 256,31 g/mol Smältpunkt: 371°C |
MDL Number:
MFCD07371389 CAS nummer: 60511-85-5 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...