1,3-Dibromo-4-nitrobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,4-Dibromo-1-nitrobenzene

D4790-1G D4790-5G
TCIAD4790-1GEA 590 SEK
TCIAD4790-1G TCIAD4790-5G
1,3-Dibromo-4-nitrobenzene ≥98.0%
1,3-Dibromo-4-nitrobenzene
Formel: C₆H₃Br₂NO₂
MW: 280,903 g/mol
MDL Number: MFCD02660416
CAS nummer: 51686-78-3

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR