4,6-Dichloro-2-methylquinoline ≥98.0%

Leverantör: TCI
Varning

D4782-1G D4782-5G
TCIAD4782-1GEA 1125 SEK
TCIAD4782-1G TCIAD4782-5G
4,6-Dichloro-2-methylquinoline ≥98.0%
4,6-Dichloro-2-methylquinoline
Formel: C₁₀H₇Cl₂N
MDL Number: MFCD00272372
CAS nummer: 53342-53-3

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR