2,6-Dichloro-4-iodopyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

D4774-1G D4774-5G
TCIAD4774-1GEA 605 SEK
TCIAD4774-1G TCIAD4774-5G
2,6-Dichloro-4-iodopyridine ≥98.0%
2,6-Dichloro-4-iodopyridine
Formel: C₅H₂CI₂IN
MW: 273,89 g/mol
MDL Number: MFCD07368400
CAS nummer: 98027-84-0

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystaline
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR