(3,3,3-Trifluoropropyl)dichloromethylsilane ≥98.0%

Leverantör: TCI
Fara

Synonyms: Dichloro(methyl)(3,3,3-trifluoropropyl)silane, (3,3,3-Trifluoropropyl)methyldichlorosilane

D4729-5G D4729-25G
TCIAD4729-5GEA 215 SEK
TCIAD4729-5G TCIAD4729-25G
(3,3,3-Trifluoropropyl)dichloromethylsilane ≥98.0%
(3,3,3-Trifluoropropyl)dichloromethylsilane
Formel: C₄H₇Cl₂F₃Si
Kokpunkt: 123°C
Flampunkt: 15°C
MDL Number: MFCD00039294
CAS nummer: 675-62-7
EINECS: 211-623-4
UN: 2985
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR