2,5-Di(2-thienyl)thieno[3,2-b]thiophene ≥94.0%
Leverantör: TCI EUROPE
Synonyms: 2,5-Di(thiophen-2-yl)thieno[3,2-b]thiophene
D4725-200MG D4725-1G
TCIAD4725-200MGEA 1225 SEK
TCIAD4725-200MG TCIAD4725-1G
2,5-Di(2-thienyl)thieno[3,2-b]thiophene ≥94.0%
2,5-Di(2-thienyl)thieno[3,2-b]thiophene
Formel: C₁₄H₈S₄
MW: 304,46 g/mol
MDL Number: MFCD11114556
CAS nummer: 21210-90-2
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 94.0 %