Diethyl diselenide ≥96.0%
Leverantör: TCI
|
Fara
|
D4715-1G
D4715-5G
TCIAD4715-1GEA
845
SEK
TCIAD4715-1G
TCIAD4715-5G
Diethyl diselenide ≥96.0%
Diethyl diselenide
Formel:
C₄H₁₀Se₂ Kokpunkt: 85°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00015491 CAS nummer: 628-39-7 EINECS: 211-042-6 UN: 3440 ADR: 6.1,III |
Specifikation testresultat
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...