2',5-Dichloro-2-hydroxybenzophenone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (5-Chloro-2-hydroxyphenyl)(2-chlorophenyl)methanone

D4703-1G D4703-5G
TCIAD4703-1GEA 415 SEK
TCIAD4703-1G TCIAD4703-5G
2',5-Dichloro-2-hydroxybenzophenone ≥98.0%
2',5-Dichloro-2-hydroxybenzophenone
Formel: C₁₃H₈Cl₂O₂
MW: 267,11 g/mol
MDL Number: MFCD02682994
CAS nummer: 61785-35-1
EINECS: 262-965-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR